Please login to your online account to display your discounted pricing

anti-CD63, Clone: MX-49.129.5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328301M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Full length CD63 of human origin
Isotype κ
Quantity 1mL
Regulatory Status RUO
Clone MX-49.129.5
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Antigen CD63
Host Species Mouse
Test Specificity This MAb recognizes protein of 26kDa-60kDa, which is identified as CD63. Its epitope is different from that of MAb LAMP3/529. The tetraspanins are integral membrane proteins expressed on cell surface and granular membranes of hematopoietic cells and are components of multi-molecular complexes with specific integrins. The tetraspanin CD63 is a lysosomal membrane glycoprotein that translocates to the plasma membrane after platelet activation. CD63 is expressed on activated platelets, monocytes and macrophages, and is weakly expressed on granulocytes, T cell and B cells. It is located on the basophilic granule membranes and on the plasma membranes of lymphocytes and granulocytes. CD63 is a member of the TM4 superfamily of leukocyte glycoproteins that includes CD9, CD37 and CD53, which contain four transmembrane regions. CD63 may play a role in phagocytic and intracellular lysosome-phagosome fusion events. CD63 deficiency is associated with Hermansky-Pudlak syndrome and is strongly expressed during the early stages of melanoma progression.
Formulation PBS with 0.05% BSA.
Gene Accession No. P08962
Primary or Secondary Primary
Gene ID (Entrez) 967
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL