Please login to your online account to display your discounted pricing

anti-CD6, Clone: C6/372, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2328271ML

 View all options for this product


For Research Use Only

Catalog No. NBP2328271M


Description & Specifications


Antigen CD6
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone C6/372
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P30203
Host Species Mouse
Immunogen Human recombinant CD6 protein
Isotype IgG1
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 923