Please login to your online account to display your discounted pricing

anti-CD6, Clone: C6/372, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232827

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232827

  • / Each
 

Description & Specifications

Specifications

Antigen CD6
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Cross Reactivity Human
Quantity 0.5mL
Immunogen Human recombinant CD6 protein
Primary or Secondary Primary
Clone C6/372
Gene Accession No. P30203
Concentration 0.2mg/mL
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Regulatory Status RUO
Gene ID (Entrez) 923
Host Species Mouse