Please login to your online account to display your discounted pricing

anti-CD6, Clone: 3F7B5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329421ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329421M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD6
Format Purified
Host Species Mouse
Isotype IgG1
Regulatory Status RUO
Clone 3F7B5
Quantity 1mL
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P30203
Gene ID (Entrez) 923
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Immunogen Human rheumatoid synovial T cell line ST-1
Primary or Secondary Primary
Concentration 0.2mg/mL