Please login to your online account to display your discounted pricing

anti-CD6, Clone: 3F7B5, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 3F7B5
Host Species Mouse
Isotype IgG1
Immunogen Human rheumatoid synovial T cell line ST-1
Antigen CD6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232942

 
novus biologicals canada
NBP232942
Unlabeled 0.5mL Each for $321.00

NBP2329421M

 
novus biologicals canada
NBP2329421ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 3F7B5
Host Species Mouse
Isotype IgG1
Immunogen Human rheumatoid synovial T cell line ST-1
Antigen CD6
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P30203
Gene ID (Entrez) 923
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL