Please login to your online account to display your discounted pricing

anti-CD5, Clone: CRIS-1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232825

 View all options for this product


For Research Use Only

Catalog No. NBP232825


Description & Specifications


Antigen CD5
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone CRIS-1
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P06127
Host Species Mouse
Immunogen Stimulated human leukocytes
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 921
Test Specificity Rhesus Monkey.