Please login to your online account to display your discounted pricing

anti-CD5, Clone: CRIS-1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone CRIS-1
Host Species Mouse
Isotype κ
Immunogen Stimulated human leukocytes
Antigen CD5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232825

 
novus biologicals canada
NBP232825
Unlabeled 0.5mL Each for $321.00

NBP2328251M

 
novus biologicals canada
NBP2328251ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone CRIS-1
Host Species Mouse
Isotype κ
Immunogen Stimulated human leukocytes
Antigen CD5
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Test Specificity Rhesus Monkey.
Gene Accession No. P06127
Gene ID (Entrez) 921
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL