Please login to your online account to display your discounted pricing

anti-CD5, Clone: CRIS-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328251ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328251M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Isotype κ
Cross Reactivity Human
Cross Reactivity Primate
Regulatory Status RUO
Format Purified
Immunogen Stimulated human leukocytes
Antigen CD5
Host Species Mouse
Clone CRIS-1
Formulation PBS with 0.05% BSA.
Gene Accession No. P06127
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 921
Test Specificity Rhesus Monkey.
Concentration 0.2mg/mL
Conjugate Unlabeled
Quantity 1mL