Please login to your online account to display your discounted pricing

anti-CD5, Clone: CD5/54/F6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232824

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232824

  • / Each
 

Description & Specifications

Specifications

Antigen CD5
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone CD5/54/F6
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P06127
Host Species Mouse
Immunogen A synthetic peptide from the intracellular region of human CD5
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 921