Please login to your online account to display your discounted pricing

anti-CD5, Clone: CD5/54/F6, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone CD5/54/F6
Host Species Mouse
Isotype κ
Immunogen A synthetic peptide from the intracellular region of human CD5
Antigen CD5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328241M

 
novus biologicals canada
NBP2328241ML
Unlabeled 1mL Each for $429.00

NBP232824

 
novus biologicals canada
NBP232824
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone CD5/54/F6
Host Species Mouse
Isotype κ
Immunogen A synthetic peptide from the intracellular region of human CD5
Antigen CD5
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P06127
Gene ID (Entrez) 921
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL