Please login to your online account to display your discounted pricing

anti-CD45RB, Clone: BRA-11/BRA-11G, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233021

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233021

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Immunogen Non-T
Isotype κ
Format Purified
Host Species Mouse
Cross Reactivity Human
Gene Accession No. P08575
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Gene ID (Entrez) 5788
Concentration 0.2mg/mL
Conjugate Unlabeled
Quantity 0.5mL
Test Specificity Reacts with Rhesus Monkey. Rat (Negative).
Clone BRA-11/BRA-11G
Antigen CD45RB