Please login to your online account to display your discounted pricing

anti-CD45RB, Clone: BRA-11/BRA-11G, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone BRA-11/BRA-11G
Host Species Mouse
Isotype κ
Immunogen Non-T
Antigen CD45RB
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233021

 
novus biologicals canada
NBP233021
Unlabeled 0.5mL Each for $321.00

NBP2330211M

 
novus biologicals canada
NBP2330211ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone BRA-11/BRA-11G
Host Species Mouse
Isotype κ
Immunogen Non-T
Antigen CD45RB
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Reacts with Rhesus Monkey. Rat (Negative).
Gene Accession No. P08575
Gene ID (Entrez) 5788
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL