Please login to your online account to display your discounted pricing

anti-CD45RB, Clone: BRA-11/BRA-11G, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2330211ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2330211M

  • / Each
 

Description & Specifications

Specifications

Antigen CD45RB
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Non-T
Isotype κ
Regulatory Status RUO
Cross Reactivity Human
Quantity 1mL
Clone BRA-11/BRA-11G
Concentration 0.2mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Gene Accession No. P08575
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Test Specificity Reacts with Rhesus Monkey. Rat (Negative).