Please login to your online account to display your discounted pricing

anti-CD45, Clone: 2B11, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2294001ML

 View all options for this product


For Research Use Only

Catalog No. NBP2294001M


Description & Specifications


Antigen CD45
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone 2B11
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Concentrated
Formulation Culture Medium
Host Species Mouse
Immunogen Neoplastic cells from Tcell lymphoma were used as the immunogen for the CD45 2B11 antibody.
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Test Specificity Recognizes a 180-185kDa protein, identified as isoform of leukocyte common antigen (CD45RO) (4th Leucocyte Typing Workshop: Code No. N31). The epitope recognized by this antibody is sensitive to neuraminidase digestion. This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas. Chimpanzee, Common Marmoset.