Please login to your online account to display your discounted pricing

anti-CD45, Clone: 2B11, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 2B11
Host Species Mouse
Isotype κ
Immunogen Neoplastic cells from Tcell lymphoma were used as the immunogen for the CD45 2B11 antibody.
Antigen CD45
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229400

 
novus biologicals canada
NBP229400
Unlabeled 0.5mL Each for $321.00

NBP2294001M

 
novus biologicals canada
NBP2294001ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 2B11
Host Species Mouse
Isotype κ
Immunogen Neoplastic cells from Tcell lymphoma were used as the immunogen for the CD45 2B11 antibody.
Antigen CD45
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Recognizes a 180-185kDa protein, identified as isoform of leukocyte common antigen (CD45RO) (4th Leucocyte Typing Workshop: Code No. N31). The epitope recognized by this antibody is sensitive to neuraminidase digestion. This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas. Chimpanzee, Common Marmoset.
Gene ID (Entrez) 5788
Formulation Culture Medium