Please login to your online account to display your discounted pricing

anti-CD4, Clone: B486A1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002181

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002181

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Primary or Secondary Primary
Concentration 1.0mg/mL
Gene Accession No. P01730
Gene ID (Entrez) 920
Conjugate Unlabeled
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Test Specificity Human. Other species have not been tested.
Isotype IgG1
Immunogen Purified recombinant fragment of human CD4 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P01730]
Quantity 0.1mL
Clone B486A1
Cross Reactivity Human
Format Precipitation
Regulatory Status RUO
Antigen CD4
Host Species Mouse