Please login to your online account to display your discounted pricing

anti-CD4, Clone: B486A1, Novus Biologicals™

$139.00 - $259.02

Specifications

Clone B486A1
Host Species Mouse
Isotype IgG1
Immunogen Purified recombinant fragment of human CD4 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P01730]
Antigen CD4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002181SS

 
novus biologicals canada
NB1002181SS
Unlabeled 0.025mL Each for $139.00

NB1002181

 
novus biologicals canada
NB1002181
Unlabeled 0.1mL Each for $259.02
Description & Specifications

Specifications

Clone B486A1
Host Species Mouse
Isotype IgG1
Immunogen Purified recombinant fragment of human CD4 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P01730]
Antigen CD4
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human. Other species have not been tested.
Gene Accession No. P01730
Gene ID (Entrez) 920
Formulation PBS
Concentration 1.0mg/mL