Please login to your online account to display your discounted pricing

anti-CD4, Clone: B486A1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002181SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002181SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 920
Conjugate Unlabeled
Gene Accession No. P01730
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Human. Other species have not been tested.
Formulation PBS
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Purified recombinant fragment of human CD4 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P01730]
Concentration 1.0mg/mL
Cross Reactivity Human
Host Species Mouse
Isotype IgG1
Antigen CD4
Quantity 0.025mL
Format Precipitation
Regulatory Status RUO
Clone B486A1