Please login to your online account to display your discounted pricing

anti-CD39/ENTPD1, Clone: 17B5F11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225223SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225223SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 953
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Test Specificity This product reacts with Human, Mouse and Rat.
Clone 17B5F11
Isotype λ
Quantity 0.025mg
Format Purified
Immunogen Amino acids 102-208 of human CD39 were used as the immunogen for the antibody.
Host Species Mouse
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen CD39/ENTPD1
Gene Accession No. NP_001191742
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Formulation CD39 antibody at 0.5mg/mL in PBS with 0.05% BSA