Please login to your online account to display your discounted pricing

anti-CD39/ENTPD1, Clone: 17B5F11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225223

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225223

  • / Each
 

Description & Specifications

Specifications

Antigen CD39/ENTPD1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 17B5F11
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation CD39 antibody at 0.5mg/mL in PBS with 0.05% BSA
Gene Accession No. NP_001191742
Host Species Murine
Immunogen Amino acids 102-208 of human CD39 were used as the immunogen for the antibody.
Isotype λ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 953
Test Specificity This product reacts with Human, Mouse and Rat.
Target Species Human
Target Species Murine
Target Species Rat