Please login to your online account to display your discounted pricing

anti-CD39/ENTPD1, Clone: 17B5F11, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 17B5F11
Host Species Murine
Isotype λ
Immunogen Amino acids 102-208 of human CD39 were used as the immunogen for the antibody.
Antigen CD39/ENTPD1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225223SS

 
novus biologicals canada
NBP225223SS
Unlabeled 0.025mg Each for $139.00

NBP225223

 
novus biologicals canada
NBP225223
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Clone 17B5F11
Host Species Murine
Isotype λ
Immunogen Amino acids 102-208 of human CD39 were used as the immunogen for the antibody.
Antigen CD39/ENTPD1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This product reacts with Human, Mouse and Rat.
Gene Accession No. NP_001191742
Gene ID (Entrez) 953
Formulation CD39 antibody at 0.5mg/mL in PBS with 0.05% BSA
Concentration 0.5mg/mL