Please login to your online account to display your discounted pricing

anti-CD34, Clone: QBEnd/10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329321ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329321M

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Primate
Quantity 1mL
Isotype κ
Host Species Mouse
Format Purified
Antigen CD34
Regulatory Status RUO
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Gene ID (Entrez) 947
Concentration 0.2mg/mL
Gene Accession No. P28906
Test Specificity Reacts with Cynomolgus Monkey, Rhesus Monkey. Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative).
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Clone QBEnd/10