Please login to your online account to display your discounted pricing

anti-CD34, Clone: QBEnd/10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232932

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232932

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Primate
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD34
Host Species Mouse
Quantity 0.5mL
Isotype κ
Regulatory Status RUO
Format Purified
Primary or Secondary Primary
Test Specificity Reacts with Cynomolgus Monkey, Rhesus Monkey. Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative).
Conjugate Unlabeled
Gene Accession No. P28906
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 947
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Clone QBEnd/10