Please login to your online account to display your discounted pricing

anti-CD34, Clone: QBEnd/10, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone QBEnd/10
Host Species Mouse
Isotype κ
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Antigen CD34
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329321M

 
novus biologicals canada
NBP2329321ML
Unlabeled 1mL Each for $429.00

NBP232932

 
novus biologicals canada
NBP232932
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone QBEnd/10
Host Species Mouse
Isotype κ
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Antigen CD34
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Reacts with Cynomolgus Monkey, Rhesus Monkey. Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative).
Gene Accession No. P28906
Gene ID (Entrez) 947
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL