Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: TLD-3A12, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10064796SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10064796T

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Activated
Host Species Mouse
Antigen CD31/PECAM-1
Isotype IgG1
Regulatory Status RUO
Clone TLD-3A12
Formulation PBS
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 5175
Cross Reactivity Rat
Test Specificity Recognizes rat PECAM-1 (CD31), a 661 amino acid type 1 transmembrane protein expressed primarily on endothelial cells, platelets and leucocytes. May cross react with Rhesus Monkey.
Concentration 1.0mg/mL
Monoclonal or Polyclonal Monoclonal
Quantity 0.05mg