Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: TLD-3A12, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10064796

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10064796

  • / Each
 

Description & Specifications

Specifications

Antigen CD31/PECAM-1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone TLD-3A12
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Rat
Format Purified
Formulation PBS
Host Species Murine
Immunogen Activated
Isotype IgG1
Quantity 0.25mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5175
Test Specificity Recognizes rat PECAM-1 (CD31), a 661 amino acid type 1 transmembrane protein expressed primarily on endothelial cells, platelets and leucocytes. May cross react with Rhesus Monkey.
Target Species Rat