Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: JC/70A, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600562

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600562

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5175
Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. P16284
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Test Specificity Human and rabbit. Rabbit data from customer review.
Format Supernatant
Immunogen Membrane prepration of a spleen from a patient with hairy cell leukemia.
Cross Reactivity Human
Cross Reactivity Rabbit
Host Species Mouse
Isotype κ
Antigen CD31/PECAM-1
Regulatory Status RUO
Quantity 0.5mL
Clone JC/70A