Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: JC/70A, Novus Biologicals™

$329.00 - $329.00

Specifications

Clone JC/70A
Host Species Mouse
Isotype κ
Immunogen Membrane prepration of a spleen from a patient with hairy cell leukemia.
Antigen CD31/PECAM-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB60056201M

 
novus biologicals canada
NB60056201ML
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone JC/70A
Host Species Mouse
Isotype κ
Immunogen Membrane prepration of a spleen from a patient with hairy cell leukemia.
Antigen CD31/PECAM-1
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and rabbit. Rabbit data from customer review.
Gene Accession No. P16284
Gene ID (Entrez) 5175