Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: CB31, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225197SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225197SS

  • / Each
 

Description & Specifications

Specifications

Antigen CD31/PECAM-1
Applications Flow Cytometry
Clone CB31
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation CD31 antibody at 0.5mg/mL in PBS containing 0.05% BSA
Gene Accession No. NP_000433
Host Species Murine
Immunogen Human umbilical vein endothelial cells were used as immunogen for this antibody.
Isotype κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5175
Test Specificity This product reacts with Human.
Target Species Human