Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: CB31, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225197

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225197

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5175
Format Purified
Formulation CD31 antibody at 0.5mg/mL in PBS containing 0.05% BSA
Host Species Mouse
Applications Flow Cytometry
Isotype κ
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product reacts with Human.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene Accession No. NP_000433
Quantity 0.1mg
Cross Reactivity Human
Antigen CD31/PECAM-1
Immunogen Human umbilical vein endothelial cells were used as immunogen for this antibody.
Clone CB31