Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: CB31, Novus Biologicals™

$139.00 - $274.00

Specifications

Clone CB31
Host Species Mouse
Isotype κ
Immunogen Human umbilical vein endothelial cells were used as immunogen for this antibody.
Antigen CD31/PECAM-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225197

 
novus biologicals canada
NBP225197
Unlabeled 0.1mg Each for $274.00

NBP225197SS

 
novus biologicals canada
NBP225197SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone CB31
Host Species Mouse
Isotype κ
Immunogen Human umbilical vein endothelial cells were used as immunogen for this antibody.
Antigen CD31/PECAM-1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Test Specificity This product reacts with Human.
Gene Accession No. NP_000433
Gene ID (Entrez) 5175
Formulation CD31 antibody at 0.5mg/mL in PBS containing 0.05% BSA
Concentration 0.5mg/mL