Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: CB31, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP226589SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP226589SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_000433
Quantity 0.025mg
Concentration 0.5mg/mL
Applications Flow Cytometry
Isotype κ
Antigen CD31/PECAM-1
Regulatory Status RUO
Cross Reactivity Human
Formulation PBS Filter sterilized. Sodium azide free
Primary or Secondary Primary
Test Specificity This product reacts with Human.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5175
Clone CB31
Conjugate Unlabeled
Format Purified
Immunogen Human umbilical vein endothelial cells were used as immunogen for this antibody.
Host Species Mouse