Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: CB31, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP226589

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP226589

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Isotype κ
Applications Flow Cytometry
Regulatory Status RUO
Antigen CD31/PECAM-1
Quantity 0.1mg
Test Specificity This product reacts with Human.
Gene ID (Entrez) 5175
Gene Accession No. NP_000433
Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation PBS Filter sterilized. Sodium azide free
Immunogen Human umbilical vein endothelial cells were used as immunogen for this antibody.
Clone CB31
Format Purified
Cross Reactivity Human
Host Species Mouse