Please login to your online account to display your discounted pricing

anti-CD28, Clone: CB28, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232928

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232928

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Isotype κ
Quantity 0.5mL
Cross Reactivity Human
Clone CB28
Antigen CD28
Immunogen Recombinant human CD28 protein
Gene Accession No. P10747
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 940
Concentration 0.2mg/mL
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Conjugate Unlabeled
Regulatory Status RUO