Please login to your online account to display your discounted pricing

anti-CD28, Clone: C28/77, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328171ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328171M

  • / Each
 

Description & Specifications

Specifications

Antigen CD28
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Clone C28/77
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P10747
Host Species Mouse
Immunogen Recombinant human CD28 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 940