Please login to your online account to display your discounted pricing

anti-CD28, Clone: C28/77, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232817

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232817

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Antigen CD28
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Isotype κ
Format Purified
Host Species Mouse
Clone C28/77
Cross Reactivity Human
Immunogen Recombinant human CD28 protein
Formulation PBS with 0.05% BSA.
Gene Accession No. P10747
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 940
Conjugate Unlabeled
Regulatory Status RUO
Concentration 0.2mg/mL