Please login to your online account to display your discounted pricing

anti-CD28, Clone: C28/75, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232816

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232816

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Regulatory Status RUO
Clone C28/75
Format Purified
Immunogen Recombinant human CD28 protein
Antigen CD28
Host Species Mouse
Isotype κ
Quantity 0.5mL
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Cross Reactivity Human
Gene Accession No. P10747
Concentration 0.2mg/mL
Gene ID (Entrez) 940