Please login to your online account to display your discounted pricing

anti-CD28, Clone: C28/75, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone C28/75
Host Species Mouse
Isotype κ
Immunogen Recombinant human CD28 protein
Antigen CD28
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232816

 
novus biologicals canada
NBP232816
Unlabeled 0.5mL Each for $321.00

NBP2328161M

 
novus biologicals canada
NBP2328161ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone C28/75
Host Species Mouse
Isotype κ
Immunogen Recombinant human CD28 protein
Antigen CD28
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Gene Accession No. P10747
Gene ID (Entrez) 940
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL