Please login to your online account to display your discounted pricing

anti-CD2, Clone: UMCD2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232815

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232815

  • / Each
 

Description & Specifications

Specifications

Antigen CD2
Format Purified
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Host Species Mouse
Cross Reactivity Human
Quantity 0.5mL
Clone UMCD2
Conjugate Unlabeled
Gene ID (Entrez) 914
Formulation PBS with 0.05% BSA.
Gene Accession No. P06729
Concentration 0.2mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Immunogen Human Thymocytes/Sezary T cells