Please login to your online account to display your discounted pricing

anti-CD2, Clone: UMCD2, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone UMCD2
Host Species Mouse
Isotype κ
Immunogen Human Thymocytes/Sezary T cells
Antigen CD2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328151M

 
novus biologicals canada
NBP2328151ML
Unlabeled 1mL Each for $429.00

NBP232815

 
novus biologicals canada
NBP232815
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone UMCD2
Host Species Mouse
Isotype κ
Immunogen Human Thymocytes/Sezary T cells
Antigen CD2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P06729
Gene ID (Entrez) 914
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL