Please login to your online account to display your discounted pricing

anti-CD2, Clone: UMCD2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328151ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328151M

  • / Each
 

Description & Specifications

Specifications

Format Purified
Clone UMCD2
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD2
Host Species Mouse
Immunogen Human Thymocytes/Sezary T cells
Isotype κ
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Gene Accession No. P06729
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 914
Conjugate Unlabeled
Concentration 0.2mg/mL
Regulatory Status RUO
Quantity 1mL