Please login to your online account to display your discounted pricing

anti-CD2, Clone: RPA-2.10, Novus Biologicals™

$109.00 - $329.00

Specifications

Clone RPA-2.10
Host Species Murine
Isotype κ
Immunogen A recombinant protein was used as the immunogen for the CD2 antibody.
Antigen CD2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225200SS

 
novus biologicals canada
NBP225200SS
Unlabeled 0.025mg Each for $109.00

NBP225200

 
novus biologicals canada
NBP225200
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Clone RPA-2.10
Host Species Murine
Isotype κ
Immunogen A recombinant protein was used as the immunogen for the CD2 antibody.
Antigen CD2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Blocking
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Neutralization
Applications Western Blot
Test Specificity This product reacts with Human.
Gene Accession No. NP_001758
Gene ID (Entrez) 914
Formulation CD2 RPA-2.10 antibody at 0.5mg/mL in PBS containing 0.05% BSA
Concentration 0.5mg/mL