Please login to your online account to display your discounted pricing

anti-CD2, Clone: BH1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232926

 View all options for this product


For Research Use Only

Catalog No. NBP232926


Description & Specifications


Antigen CD2
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone BH1
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P06729
Host Species Mouse
Immunogen Human CD2 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 914