Please login to your online account to display your discounted pricing

anti-CD2, Clone: BH1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone BH1
Host Species Mouse
Isotype κ
Immunogen Human CD2 protein
Antigen CD2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232926

 
novus biologicals canada
NBP232926
Unlabeled 0.5mL Each for $321.00

NBP2329261M

 
novus biologicals canada
NBP2329261ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone BH1
Host Species Mouse
Isotype κ
Immunogen Human CD2 protein
Antigen CD2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene Accession No. P06729
Gene ID (Entrez) 914
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL