Please login to your online account to display your discounted pricing

anti-CD2, Clone: BH1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329261ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329261M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Regulatory Status RUO
Isotype κ
Host Species Mouse
Clone BH1
Immunogen Human CD2 protein
Quantity 1mL
Format Purified
Antigen CD2
Concentration 0.2mg/mL
Gene Accession No. P06729
Gene ID (Entrez) 914
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Cross Reactivity Human