Please login to your online account to display your discounted pricing

anti-CD2, Clone: 1E7E8.G4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232814

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232814

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Format Purified
Host Species Mouse
Quantity 0.5mL
Immunogen Human Thymocytes/Sezary T cells
Regulatory Status RUO
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. P06729
Primary or Secondary Primary
Isotype κ
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 914
Clone 1E7E8.G4
Antigen CD2