Please login to your online account to display your discounted pricing

anti-CD1b, Clone: RIV12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232925

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232925

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Isotype κ
Format Purified
Antigen CD1b
Regulatory Status RUO
Host Species Mouse
Cross Reactivity Human
Quantity 0.5mL
Clone RIV12
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 910
Gene Accession No. P29016
Immunogen Human peripheral lymphocytes
Concentration 0.2mg/mL