Please login to your online account to display your discounted pricing

anti-CD1b, Clone: RIV12, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone RIV12
Host Species Mouse
Isotype κ
Immunogen Human peripheral lymphocytes
Antigen CD1b
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232925

 
novus biologicals canada
NBP232925
Unlabeled 0.5mL Each for $321.00

NBP2329251M

 
novus biologicals canada
NBP2329251ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone RIV12
Host Species Mouse
Isotype κ
Immunogen Human peripheral lymphocytes
Antigen CD1b
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene Accession No. P29016
Gene ID (Entrez) 910
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL