Please login to your online account to display your discounted pricing

anti-CD1a, Clone: C1A/711, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233018

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233018

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human CD1a protein
Regulatory Status RUO
Quantity 0.5mL
Isotype κ
Clone C1A/711
Cross Reactivity Human
Gene Accession No. P06126
Primary or Secondary Primary
Gene ID (Entrez) 909
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Host Species Mouse
Antigen CD1a
Format Purified