Please login to your online account to display your discounted pricing

anti-CD1a, Clone: C1A/711, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone C1A/711
Host Species Mouse
Isotype κ
Immunogen Recombinant human CD1a protein
Antigen CD1a
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233018

 
novus biologicals canada
NBP233018
Unlabeled 0.5mL Each for $321.00

NBP2330181M

 
novus biologicals canada
NBP2330181ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone C1A/711
Host Species Mouse
Isotype κ
Immunogen Recombinant human CD1a protein
Antigen CD1a
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P06126
Gene ID (Entrez) 909
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL